APO455828113 Apollo Scientific Ltd

1-Amino-5-bromoisoquinoline

  • MDL number MFCD11226920
  • Cas Number : 852570-80-0
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-1-isoquinolinylamine
Inchi Code 1S/C9H7BrN2/c10-8-3-1-2-7-6(8)4-5-12-9(7)11/h1-5H,(H2,11,12)
Inchi Key ATENCABMFYJWGZ-UHFFFAOYSA-N

Description

Synonym 5-Bromoisoquinolin-1-amine, 1-Amino-5-bromo-2-azanaphthalene
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-amino-5-bromoisoquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-amino-5-bromoisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart