APO455828114 Apollo Scientific Ltd

2-Bromo-4-iodoaniline

  • MDL number MFCD07780668
  • Cas Number : 29632-73-3
  • Molecular Weight : 297.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-85
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-iodoaniline

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-iodoaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart