APO455828116 Apollo Scientific Ltd

2-Bromo-6-methyl-4-nitrophenol

  • MDL number MFCD26392591
  • Cas Number : 4186-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-122
Synonym 6-Bromo-4-nitro-o-cresol, 3-Bromo-2-hydroxy-5-nitrotoluene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=62394

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=62394
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart