APO455828119 Apollo Scientific Ltd

4-Amino-3-hydroxybenzonitrile

  • MDL number MFCD06657033
  • Cas Number : 55586-26-0
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-3-hydroxybenzonitrile
Inchi Code 1S/C7H6N2O/c8-4-5-1-2-6(9)7(10)3-5/h1-3,10H,9H2
Inchi Key HKZPOGZBKIWMPO-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-hydroxybenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-hydroxybenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart