APO455828123 Apollo Scientific Ltd

Quinoxaline-6-carboxaldehyde

  • MDL number MFCD00805834
  • Cas Number : 130345-50-5
  • Molecular Weight : 158.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 130-131
Iupac Name 6-quinoxalinecarbaldehyde
Inchi Code 1S/C9H6N2O/c12-6-7-1-2-8-9(5-7)11-4-3-10-8/h1-6H
Inchi Key UGOIXUFOAODGNI-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 130-131
Product Link https://store.apolloscientific.co.uk/product/quinoxaline-6-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/quinoxaline-6-carboxaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart