APO455828124 Apollo Scientific Ltd

5-Cyanopyridine-3-boronic acid

  • MDL number MFCD07374882
  • Cas Number : 497147-93-0
  • Molecular Weight : 147.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-145
Synonym 3-Cyanopyridine-5-boronic acid, 3-Borono-5-cyanopyridine
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-cyanopyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-cyanopyridine-3-boronic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart