APO455828126 Apollo Scientific Ltd

N-(4,6-Dimethylpyrimidin-2-yl)-N-methylbenzene-1,4-diamine

  • MDL number MFCD00202978
  • Cas Number : 220844-79-1
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-136
Synonym 2-[(4-Aminophenyl)(methyl)amino]-4,6-dimethylpyrimidine, 2-[(4-Aminophenyl)(methyl)amino]-4,6-dimethyl-1,3-diazine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-46-dimethylpyrimidin-2-yl-n-methylbenzene-14-diamine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/n-46-dimethylpyrimidin-2-yl-n-methylbenzene-14-diamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart