APO455828132 Apollo Scientific Ltd

3-Bromo-5-methoxybenzonitrile

  • MDL number MFCD08061971
  • Cas Number : 867366-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Synonym 3-Bromo-5-cyanoanisole, 5-Bromo-m-anisonitrile
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-methoxybenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-methoxybenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart