APO455828134 Apollo Scientific Ltd

2,4-Dichloro-6-iodoaniline

  • MDL number MFCD08533279
  • Cas Number : 697-90-5
  • Molecular Weight : 287.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-81
Synonym 1-Amino-2,4-dichloro-6-iodobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dichloro-6-iodoaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-dichloro-6-iodoaniline
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart