APO455828136 Apollo Scientific Ltd

Methyl 5-methyl-2-nitrobenzoate

  • MDL number MFCD00157275
  • Cas Number : 20587-30-8
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-methyl-2-nitrobenzoate
Inchi Code 1S/C9H9NO4/c1-6-3-4-8(10(12)13)7(5-6)9(11)14-2/h3-5H,1-2H3
Inchi Key KFOICDVZQKFCGM-UHFFFAOYSA-N

Description

Synonym 2-(Methoxycarbonyl)-4-methylnitrobenzene, 3-(Methoxycarbonyl)-4-nitrotoluene
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-5-methyl-2-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-methyl-2-nitrobenzoate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart