APO455828138 Apollo Scientific Ltd

Pyrimidine-2-carboxaldehyde

  • MDL number MFCD07778416
  • Cas Number : 27427-92-5
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Formylpyrimidine, 2-Formyl-1,3-diazine
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/pyrimidine-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/pyrimidine-2-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart