APO455828154 Apollo Scientific Ltd

5-Bromo-3-chloropyridine-2-carbonitrile

  • MDL number MFCD12827550
  • Cas Number : 945557-04-0
  • Molecular Weight : 217.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-3-chloro-2-pyridinecarbonitrile
Inchi Code 1S/C6H2BrClN2/c7-4-1-5(8)6(2-9)10-3-4/h1,3H
Inchi Key SIYPZKHSXDFDRA-UHFFFAOYSA-N

Description

Synonym 5-Bromo-3-chloro-2-cyanopyridine, 5-Bromo-3-chloropicolinonitrile
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-chloropyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-chloropyridine-2-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart