APO455828156 Apollo Scientific Ltd

5-(Hydroxymethyl)pyrimidine

  • MDL number MFCD03789623
  • Cas Number : 25193-95-7
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Pyrimidin-5-yl)methanol
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-hydroxymethylpyrimidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxymethylpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart