APO455828157 Apollo Scientific Ltd

beta-Aminophenethyl alcohol

  • MDL number MFCD00130145
  • Cas Number : 7568-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-58
Synonym 2-Amino-2-phenylethan-1-ol, 2-Phenylglycinol, alpha-(Hydroxymethyl)benzylamine
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 261
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46543

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46543
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart