APO455828159 Apollo Scientific Ltd

Benzo[b]furan-5-boronic acid, pinacol ester

  • MDL number MFCD04115374
  • Cas Number : 519054-55-8
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 63-66
Iupac Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran
Inchi Code 1S/C14H17BO3/c1-13(2)14(3,4)18-15(17-13)11-5-6-12-10(9-11)7-8-16-12/h5-9H,1-4H3
Inchi Key ZQCCCOMKYKVFFN-UHFFFAOYSA-N

Description

Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran
Flash Point 90-120/0mmHg
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 63-66
Product Link https://store.apolloscientific.co.uk/product/benzobfuran-5-boronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/benzobfuran-5-boronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart