APO455828163 Apollo Scientific Ltd

5-Iodoindan-1-one

  • MDL number MFCD09744086
  • Cas Number : 511533-38-3
  • Molecular Weight : 258.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Dihydro-5-iodo-1H-inden-1-one, 2,3-Dihydro-5-iodo-1-oxo-1H-indene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-iodoindan-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-iodoindan-1-one
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart