APO455828166 Apollo Scientific Ltd

Furo[3,2-b]pyridine

  • MDL number MFCD11870744
  • Cas Number : 272-62-8
  • Molecular Weight : 119.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name furo[3,2-b]pyridine
Inchi Code 1S/C7H5NO/c1-2-7-6(8-4-1)3-5-9-7/h1-5H
Inchi Key YRTCKZIKGWZNCU-UHFFFAOYSA-N

Description

Synonym 4-Azabenzo[b]furan, 4-Aza-1-benzofuran
Flash Point 79-80/12mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/furo32-bpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/furo32-bpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart