APO455828168 Apollo Scientific Ltd

2-Amino-4-iodo-1,3-benzothiazole

  • MDL number MFCD11194508
  • Cas Number : 1039326-79-8
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Iodo-1,3-benzothiazol-2-amine
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-iodo-13-benzothiazole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-iodo-13-benzothiazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart