APO455828175 Apollo Scientific Ltd

2-(1,3-Benzoxazol-2-yl)-1-(4-tert-butylphenyl)ethan-1-one

  • MDL number MFCD04037909
  • Cas Number : 849021-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-13-benzoxazol-2-yl-1-4-tert-butylphenylethan-1-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-13-benzoxazol-2-yl-1-4-tert-butylphenylethan-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart