APO455828179 Apollo Scientific Ltd

3-Methyl-2-nitrophenylacetic acid

  • MDL number MFCD08276006
  • Cas Number : 18710-86-6
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Carboxymethyl)-2-nitrotoluene, 2-(Carboxymethyl)-6-methylnitrobenzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methyl-2-nitrophenylacetic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-2-nitrophenylacetic-acid
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart