APO455828181 Apollo Scientific Ltd

2-Benzylacrylic acid

  • MDL number MFCD00854174
  • Cas Number : 5669-19-2
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 70
Iupac Name 2-benzylacrylic acid
Inchi Code 1S/C10H10O2/c1-8(10(11)12)7-9-5-3-2-4-6-9/h2-6H,1,7H2,(H,11,12)
Inchi Key RYNDYESLUKWOEE-UHFFFAOYSA-N

Description

Synonym 2-Benzylprop-2-enoic acid, 2-(Phenylmethyl)prop-2-enoic acid
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 70
Product Link https://store.apolloscientific.co.uk/product/2-benzylacrylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-benzylacrylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart