APO455828183 Apollo Scientific Ltd

2-Chloro-4-methylpyrimidine-5-carboxylic acid

  • MDL number MFCD09863167
  • Cas Number : 188781-10-4
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Carboxy-2-chloro-4-methylpyrimidine, 2-Chloro-4-methyl-1,3-diazine-5-carboxylic acid
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-methylpyrimidine-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-methylpyrimidine-5-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart