APO455828186 Apollo Scientific Ltd

Pyrimidine-4-boronic acid

  • MDL number MFCD08234611
  • Cas Number : 852362-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Boronopyrimidine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47367

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47367
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart