APO455828191 Apollo Scientific Ltd

5-Bromopyrazine-2-carbonitrile

  • MDL number MFCD09909652
  • Cas Number : 221295-04-1
  • Molecular Weight : 183.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-pyrazinecarbonitrile
Inchi Code 1S/C5H2BrN3/c6-5-3-8-4(1-7)2-9-5/h2-3H
Inchi Key ZPBNABIKZKTQQH-UHFFFAOYSA-N

Description

Synonym 2-Bromo-5-cyanopyrazine, 2-Bromo-5-cyano-1,4-diazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromopyrazine-2-carbonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromopyrazine-2-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart