APO455828194 Apollo Scientific Ltd

5-Bromo-4-methyl-1,3-thiazole

  • MDL number MFCD12026322
  • Cas Number : 111600-83-0
  • Molecular Weight : 178.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-methyl-1,3-thiazole
Inchi Code 1S/C4H4BrNS/c1-3-4(5)7-2-6-3/h2H,1H3
Inchi Key IIMLZWMRQNCPTM-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-4-methyl-13-thiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-4-methyl-13-thiazole
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart