APO455828204 Apollo Scientific Ltd

5-Amino-1H-imidazole-4-carboxamide hydrochloride

  • MDL number MFCD00799200
  • Cas Number : 72-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-252
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-amino-1h-imidazole-4-carboxamide-hydrochloride_OR18276

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1h-imidazole-4-carboxamide-hydrochloride_OR18276
pictograms GHS07
hstatements H319;H315;H335
Availability details for
Quantity
  • Availability
  • Add to Cart