APO455828212 Apollo Scientific Ltd

[(Pyridin-4-yl)thio]acetic acid

  • MDL number MFCD00006424
  • Cas Number : 10351-19-6
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 251-260(dec.)
Iupac Name (4-pyridinylsulfanyl)acetic acid
Inchi Code 1S/C7H7NO2S/c9-7(10)5-11-6-1-3-8-4-2-6/h1-4H,5H2,(H,9,10)
Inchi Key PGUPJAPHYIEKLT-UHFFFAOYSA-N

Description

Synonym [(Pyridin-4-yl)sulphanyl]acetic acid, 4-[(Carboxymethyl)thio]pyridine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 251-260(dec.)
Product Link https://store.apolloscientific.co.uk/product/pyridin-4-ylthioacetic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/pyridin-4-ylthioacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart