APO455828235 Apollo Scientific Ltd

Isopropyl phenyl sulphide

  • MDL number MFCD00039842
  • Cas Number : 3019-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Isopropylthio)benzene, (Isopropylsulphanyl)benzene
Purity 97
Density 0.9852
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 105/ 40mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27611

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27611
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart