APO455828247 Apollo Scientific Ltd

1,4-Diethoxybenzene

  • MDL number MFCD00015146
  • Cas Number : 122-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-72
Synonym Hydroquinone diethyl ether
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 245-248
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27624

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27624
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart