APO455828288 Apollo Scientific Ltd

4-(Diethylamino)benzoic acid

  • MDL number MFCD00002538
  • Cas Number : 5429-28-7
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-195
Synonym 4-Carboxy-N,N-diethylaniline
Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-diethylaminobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-diethylaminobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart