APO455828300 Apollo Scientific Ltd

2,5-Dibromophenol

  • MDL number MFCD01851373
  • Cas Number : 28165-52-8
  • Molecular Weight : 251.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/25-dibromophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/25-dibromophenol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart