APO455828313 Apollo Scientific Ltd

1H-Benzimidazole-2-carboxaldehyde

  • MDL number MFCD00228045
  • Cas Number : 3314-30-5
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-benzimidazole-2-carbaldehyde
Inchi Code 1S/C8H6N2O/c11-5-8-9-6-3-1-2-4-7(6)10-8/h1-5H,(H,9,10)
Inchi Key DQOSJWYZDQIMGM-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1h-benzimidazole-2-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1h-benzimidazole-2-carboxaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart