APO455828315 Apollo Scientific Ltd

3-[2-(Morpholin-4-yl)ethoxy]benzaldehyde hydrochloride

  • MDL number MFCD13186601
  • Cas Number : 1609402-82-5
  • Molecular Weight : 271.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[2-(3-Formylphenoxy)ethyl]morpholine hydrochloride
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-2-morpholin-4-ylethoxybenzaldehyde-hydrochloride

Safety Information

signalword Danger
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/3-2-morpholin-4-ylethoxybenzaldehyde-hydrochloride
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart