APO455828329 Apollo Scientific Ltd

3-Acetoxy-4-methoxybenzeneboronic acid, pinacol ester

  • MDL number MFCD06795645
  • Cas Number : 917757-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(3-Acetoxy-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=46483

Safety Information

signalword Warning
pcodes P280;P309+P311
MSDS http://www.apolloscientific.co.uk/display_item.php?id=46483
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart