APO455828330 Apollo Scientific Ltd

5-Amino-3-(tert-butyl)isoxazole

  • MDL number MFCD05266697
  • Cas Number : 59669-59-9
  • Molecular Weight : 140.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(tert-Butyl)isoxazol-5-amine, 5-Amino-3-(tert-butyl)-1,2-oxazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-3-tert-butylisoxazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-3-tert-butylisoxazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart