APO455828338 Apollo Scientific Ltd

1-(Diphenylmethyl)-3-hydroxyazetidine hydrochloride

  • MDL number MFCD00209447
  • Cas Number : 90604-02-7
  • Molecular Weight : 275.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzhydryl-3-azetidinol hydrochloride
Inchi Code 1S/C16H17NO.ClH/c18-15-11-17(12-15)16(13-7-3-1-4-8-13)14-9-5-2-6-10-14;/h1-10,15-16,18H,11-12H2;1H
Inchi Key LCHTWRWPHBRTNO-UHFFFAOYSA-N

Description

Synonym 1-Benzhydrylazetidin-3-ol hydrochloride
Purity 95%
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-diphenylmethyl-3-hydroxyazetidine-hydrochloride

Safety Information

signalword Danger
pcodes P260;P270;P280
MSDS https://store.apolloscientific.co.uk/product/1-diphenylmethyl-3-hydroxyazetidine-hydrochloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart