APO455828351 Apollo Scientific Ltd

2-Amino-4-chloro-1,3-thiazole-5-carboxaldehyde, 2-BOC protected

  • MDL number MFCD18836516
  • Cas Number : 388594-31-8
  • Molecular Weight : 262.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl (4-chloro-5-formyl-1,3-thiazol-2-yl)carbamate, 2-[(tert-Butoxycarbonyl)amino]-4-chloro-5-formyl-1,3-thiazole
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-chloro-13-thiazole-5-carboxaldehyde-2-boc-protected

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-chloro-13-thiazole-5-carboxaldehyde-2-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart