APO455828352 Apollo Scientific Ltd

2-Amino-4-chloro-1,3-thiazole-5-carboxylic acid, 2-BOC protected

  • MDL number MFCD24476508
  • Cas Number : 612541-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl (5-carboxy-4-chloro-1,3-thiazol-2-yl)carbamate, 2-[(tert-Butoxycarbonyl)amino]-4-chloro-1,3-thiazole-5-carboxylic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63454

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63454
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart