APO455828353 Apollo Scientific Ltd

3-Methyl-2,4,5-tribromothiophene

  • MDL number MFCD00130086
  • Cas Number : 67869-13-0
  • Molecular Weight : 334.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31
Synonym 2,3,5-Tribromo-4-methylthiophene
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methyl-245-tribromothiophene

Safety Information

pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-245-tribromothiophene
Availability details for
Quantity
  • Availability
  • Add to Cart