APO455828356 Apollo Scientific Ltd

3-Bromo-4-methoxy-5-methylaniline

  • MDL number MFCD16618926
  • Cas Number : 1215205-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Amino-2-bromo-6-methylanisole, 5-Amino-3-bromo-2-methoxytoluene, 3-Bromo-5-methyl-p-anisidine, 5-Bromo-4-methoxy-m-toluidine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63868

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63868
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart