APO455828364 Apollo Scientific Ltd

3,5-Dichloropyridine-2-carboxylic acid

  • MDL number MFCD03230005
  • Cas Number : 81719-53-1
  • Molecular Weight : 192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 156(dec.)
Iupac Name 3,5-dichloro-2-pyridinecarboxylic acid
Inchi Code 1S/C6H3Cl2NO2/c7-3-1-4(8)5(6(10)11)9-2-3/h1-2H,(H,10,11)
Inchi Key JYKZCYLZWZCQFP-UHFFFAOYSA-N

Description

Synonym 3,5-Dichloropicolinic acid, 2-Carboxy-3,5-dichloropyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 156(dec.)
Product Link https://store.apolloscientific.co.uk/product/35-dichloropyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloropyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart