APO455828366 Apollo Scientific Ltd

2-Hydroxyisonicotinic acid

  • MDL number MFCD00955759
  • Cas Number : 22282-72-0
  • Molecular Weight : 139.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 2-Hydroxypyridine-4-carboxylic acid, 1,2-Dihydro-2-oxopyridine-4-carboxylic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxyisonicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxyisonicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart