APO455828373 Apollo Scientific Ltd

1-Methyl-1H-indole-2-boronic acid

  • MDL number MFCD01114668
  • Cas Number : 191162-40-0
  • Molecular Weight : 174.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 108-112
Iupac Name 1-methyl-1H-indol-2-ylboronic acid
Inchi Code 1S/C9H10BNO2/c1-11-8-5-3-2-4-7(8)6-9(11)10(12)13/h2-6,12-13H,1H3
Inchi Key CBPBJUTWVXLSER-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at -20
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 108-112
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-indole-2-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-indole-2-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart