APO455828376 Apollo Scientific Ltd

3-Chloro-4-(methoxycarbonyl)benzeneboronic acid

  • MDL number MFCD06659874
  • Cas Number : 603122-82-3
  • Molecular Weight : 214.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-178
Synonym Methyl 4-borono-2-chlorobenzoate
Purity 96
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-methoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-methoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart