APO455828378 Apollo Scientific Ltd

4-Nitrophenylhydrazine hydrochloride

  • MDL number MFCD00040592
  • Cas Number : 636-99-7
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 198-200(dec.)
Iupac Name 1-(4-nitrophenyl)hydrazine hydrochloride
Inchi Code 1S/C6H7N3O2.ClH/c7-8-5-1-3-6(4-2-5)9(10)11;/h1-4,8H,7H2;1H
Inchi Key ZWWXDCOPVYATOQ-UHFFFAOYSA-N

Description

Synonym 4-Hydrazinonitrobenzene hydrochloride, 1-Hydrazino-4-nitrobenzene hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 198-200(dec.)
Product Link https://store.apolloscientific.co.uk/product/4-nitrophenylhydrazine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-nitrophenylhydrazine-hydrochloride
pictograms GHS07
hstatements H302;H312;H315;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart