APO455828379 Apollo Scientific Ltd

2-Nitrophenylhydrazine hydrochloride

  • MDL number MFCD00060205
  • Cas Number : 6293-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Hydrazinonitrobenzene hydrochloride
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30263

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30263
pictograms GHS02;GHS07
hstatements H228;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart