APO455828386 Apollo Scientific Ltd

2,6-Dibromobenzo[b]furan-3(2H)-one

  • MDL number MFCD28096609
  • Cas Number : 1823370-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,6-Dibromo-1-benzofuran-3(2H)-one, 2,6-Dibromo-2,3-dihydro-3-oxobenzo[b]furan
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63952

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=63952
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart