APO455828388 Apollo Scientific Ltd

2,5-Dibromobenzo[b]furan-3(2H)-one

  • MDL number MFCD28096610
  • Cas Number : 1823918-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,5-Dibromo-1-benzofuran-3(2H)-one, 2,5-Dibromo-2,3-dihydro-3-oxobenzo[b]furan
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63953

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63953
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart