APO455828396 Apollo Scientific Ltd

2-Amino-5-bromo-4-chloropyridine

  • MDL number MFCD12407282
  • Cas Number : 942947-94-6
  • Molecular Weight : 207.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-147
Synonym 5-Bromo-4-chloropyridin-2-amine
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-bromo-4-chloropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-bromo-4-chloropyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart