APO455828399 Apollo Scientific Ltd

3-Allylanisole

  • MDL number MFCD06201195
  • Cas Number : 24743-14-4
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Prop-2-en-1-yl)anisole, 3-(3-Methoxyphenyl)prop-1-ene, 1-Methoxy-3-(prop-2-en-1-yl)benzene, Methyl 3-(prop-2-en-1-yl)phenyl ether
Purity 95
Density 0.978
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 213-216
Product Link https://store.apolloscientific.co.uk/product/3-allylanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-allylanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart